missing translation for 'onlineSavingsMsg'
Learn More

Thionin acetate

Product Code. 11355239
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355239 25 g 25g
11345239 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355239 Supplier Thermo Scientific Alfa Aesar Supplier No. A18912.14
  • ONLINE EXCLUSIVE PRICE
    £173.40
    £142.65 / 25g
    Save £30.75
    17% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a biogical stain | CAS: 78338-22-4 | C14H13N3O2S | 287.337 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Thionin acetate is a strongly staining metachromatic dye that is widely used for biological staining. Thionine can also be used in place of Schiff reagent in quantitative Feulgen staining of DNA. It can also be used to mediate electron transfer in microbial fuel cells. It is also employed as a general nuclear stain for chromatin and mucin; for staining Nissl granules by end-point buffering and for showing malignant cells in frozen biopsy sections; and as a stain for semi-thin sections of plant material for light microscopy.

Solubility
Soluble in water (∼2.5 mg/ml at 25°C), and HOAc: H2O (1:1) (1 mg/ml). Insoluble in ethanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78338-22-4
Molecular Formula C14H13N3O2S
Molecular Weight (g/mol) 287.337
MDL Number MFCD00081194
InChI Key JOIRQYHDJINFGA-UHFFFAOYSA-N
PubChem CID 2724414
IUPAC Name (7-aminophenothiazin-3-ylidene)azanium;acetate
SMILES CC(=O)[O-].C1=CC2=C(C=C1N)SC3=CC(=[NH2+])C=CC3=N2

Specifications

Solubility Information Soluble in water (∽2.5mg/ml at 25°C),and HOAc: H2O (1:1) (1mg/ml). Insoluble in ethanol.
Quantity 25 g
Formula Weight 287.34
Chemical Name or Material Thionin acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.