missing translation for 'onlineSavingsMsg'
Learn More

Thionin Acetate Salt, TRC

Product Code. 30456953
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456953 5 g 5g
30456161 2.5 g 2.50g
30450389 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456953 Supplier Toronto Research Chemicals Supplier No. TRCT3677555G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78338-22-4
Molecular Formula C12H10N3S •C2H3O2
Molecular Weight (g/mol) 287.33
Synonym 3,7-Diaminophenothiazin-5-ium Acetate, Thionin Acetate, Thionine Acetate, 3,7-Diamino-phenothiazin-5-ium Acetate
IUPAC Name 3,7-diaminophenothiazin-5-ium acetate
SMILES O=C(C)[O-].NC1=CC=C2N=C3C=CC(N)=CC3=[S+]C2=C1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H10N3S.C2H4O2/c13-7-1-3-9-11(5-7)16-12-6-8(14)2-4-10(12)15-9;1-2(3)4/h1-6H,13-14H2;1H3,(H,3,4)/q+1;/p-1
Formula Weight 287.07
Chemical Name or Material Thionin Acetate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.