missing translation for 'onlineSavingsMsg'
Learn More

Thionin acetate, MedChemExpress

Product Code. 16462429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16462429 Supplier Medchem Express Supplier No. HYD0955
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Thionin acetate (Thionine acetate) is a metachromic cationic histology dye used in biological staining widely.

Thionin acetate (Thionine acetate) is a metachromic cationic histology dye used in biological staining widely.

Specifications

Chemical Name or Material Thionin acetate
CAS 78338-22-4
Purity Grade Notes Research
Molecular Formula C14H13N3O2S
Synonym Thionine acetate
Solubility Information DMSO : 41.67 mg/mL (145.02 mM; ultrasonic and warming and heat to 60°C)
SMILES NC1=CC2=[S+]C3=C(C=CC(N)=C3)N=C2C=C1.CC([O-])=O
Molecular Weight (g/mol) 287.34
Formula Weight 287.34
Grade Research
Color Dark Green
Quantity 1 g
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.