missing translation for 'onlineSavingsMsg'
Learn More

Thiomorpholine, 98%

Product Code. 11478516
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478516 100 g 100g
11458516 5 g 5g
11468516 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478516 Supplier Thermo Scientific Alfa Aesar Supplier No. A15958.22
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-90-0 | C4H9NS | 103.183 g/mol

Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiomorpholine is used as an intermediate for blonanserin. It is used in the preparation of pyrrole derivatives, which acts as a antimycobacterial agent. It is further involved in the hypocholesterolemic activity.

Solubility
Miscible with waterMiscible with water and organic solvents.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-90-0
Molecular Formula C4H9NS
Molecular Weight (g/mol) 103.183
MDL Number MFCD00005974
InChI Key BRNULMACUQOKMR-UHFFFAOYSA-N
Synonym thiamorpholine, thiazolidinane, parathiazan, 4-thiomorpholine, tetrahydro-1,4-thiazine, 1,4-thiazane, 1,4-thiazan, 1-thia-4-azacyclohexane, unii-3a8r61g6qv, 1,4-thiazaperhydroine
PubChem CID 67164
ChEBI CHEBI:36392
IUPAC Name thiomorpholine
SMILES C1CSCCN1

Specifications

Density 1.086
Boiling Point 175°C to 177°C
Flash Point 67°C (153°F)
Refractive Index 1.5384
Quantity 100 g
UN Number UN3267
Beilstein 102550
Sensitivity Air Sensitive
Merck Index 14,9300
Solubility Information Miscible with waterMiscible with water and organic solvents.
Formula Weight 103.19
Percent Purity 98%
Chemical Name or Material Thiomorpholine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.