missing translation for 'onlineSavingsMsg'
Learn More

Thiomorpholine 1,1-Dioxide, TRC

Product Code. 30456992
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30456992 100 mg 100mg
30449241 50 mg -
30454093 10 mg -
3 options
This item is not returnable. View return policy

Product Code. 30456992

Brand: Toronto Research Chemicals TRCT344568100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39093-93-1
Formula Weight 135.0354
InChI Formula InChI=1S/C4H9NO2S/c6-8(7)3-1-5-2-4-8/h5H,1-4H2
IUPAC Name 1,4-thiazinane 1,1-dioxide
Molecular Formula C4H9NO2S
Molecular Weight (g/mol) 135.18
SMILES O=S1(=O)CCNCC1
Synonym 1,1-Dioxidothiomorpholine, 1,1-Dioxo-1-thiomorpholine, 1,1-Dioxothiomorpholine, 1,4-Thiazinane 1,1-Dioxide, Tetrahydro-1,4-thiazine 1,1-Dioxide, Tetrahydro-2H-1,4-thiazine 1,1-Dioxide, Thiomorpholine S,S-Dioxide, Thiomorpholine Dioxide
Chemical Name or Material Thiomorpholine 1,1-Dioxide

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.