missing translation for 'onlineSavingsMsg'
Learn More

Thiometon Sulfoxide, TRC

Product Code. 30453545
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30453545

Brand: Toronto Research Chemicals TRCT344675100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiometon-sulfoxide
CAS 2703-37-9
Purity Grade Notes HPLC
Molecular Formula C6 H15 O3 P S3
Quantity 100 mg
Synonym Intrathion sulfoxide, Intration sulfoxide, O,O-Dimethyl S-(2-ethylsulfinyl)ethyl dithiophosphate, O,O-Dimethyl S-(2-ethylsulfinylethyl) phosphorodithioate, Oxythiometon, Thiometon sulfoxide, Thiometon-sulfoxide
InChI Formula InChI=1S/C6H15O3PS3/c1-4-13(7)6-5-12-10(11,8-2)9-3/h4-6H2,1-3H3
SMILES CCS(=O)CCSP(=S)(OC)OC
IUPAC Name 2-ethylsulfinylethylsulfanyl-dimethoxy-sulfanylidene-λ^{5}-phosphane
Molecular Weight (g/mol) 262.35
Formula Weight 261.9921
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.