missing translation for 'onlineSavingsMsg'
Learn More

Thiomalic acid, TRC

Product Code. 30414706
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414706 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414706 Supplier Toronto Research Chemicals Supplier No. TRCT14051010G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiomalic acid
CAS 70-49-5
Molecular Formula C4H6O4S
Quantity 10 g
Synonym 2-mercapto-Butanedioic acid, mercapto-Butanedioic acid, mercapto-Succinic acid, 2-Mercaptobutanedioic acid, (±)-Mercaptosuccinic acid, 2-Mercaptosuccinic acid, 2-Thiomalic acid, DL-Mercaptosuccinic acid, DL-Thiomalic acid, Mercaptobutanedioic acid, Mercaptosuccinic acid, Monomercaptosuccinic acid, NSC 7302, alpha-Mercaptosuccinic acid
InChI Formula InChI=1S/C4H6O4S/c5-3(6)1-2(9)4(7)8/h2,9H,1H2,(H,5,6)(H,7,8)
SMILES OC(=O)CC(S)C(=O)O
IUPAC Name 2-sulfanylbutanedioic acid
Molecular Weight (g/mol) 150.15
Formula Weight 149.9987
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.