missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioisonicotinamide, 97%, Thermo Scientific™
£23.10 - £89.20
Chemical Identifiers
CAS | 2196-13-6 |
---|---|
Molecular Formula | C6H6N2S |
Molecular Weight (g/mol) | 138.19 |
MDL Number | MFCD00006437 |
InChI Key | KPIIGXWUNXGGCP-UHFFFAOYSA-N |
Synonym | thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio |
PubChem CID | 2723788 |
IUPAC Name | pyridine-4-carbothioamide |
SMILES | C1=CN=CC=C1C(=S)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10882631
|
Thermo Scientific Acros
138770050 |
5 g | Glass bottle | ||||||
11395116
|
Thermo Scientific Acros
138770250 |
25 g | Glass bottle |
£89.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
2196-13-6 | |
138.19 | |
KPIIGXWUNXGGCP-UHFFFAOYSA-N | |
2723788 | |
C1=CN=CC=C1C(=S)N |
C6H6N2S | |
MFCD00006437 | |
thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio | |
pyridine-4-carbothioamide |
Specifications
2196-13-6 | |
Brown-Yellow to Cream | |
96% min. (HPLC) | |
MFCD00006437 | |
KPIIGXWUNXGGCP-UHFFFAOYSA-N | |
pyridine-4-carbothioamide | |
2723788 | |
97% | |
Thioisonicotinamide |
198.0°C to 202.0°C | |
Authentic | |
C6H6N2S | |
thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio | |
C1=CN=CC=C1C(=S)N | |
138.19 | |
138.19 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 218-592-6
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Thioisonicotinamide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only