Learn More
Thioglycolic Acid, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical T/1000/PB05
34.60 GBP valid until 2021-02-28
Use promo code "14802" to get your promotional price.
Code : 21
Additional Details : CAS Number : 68-11-1 Weight : 0.11000kg Transport : UN number : 1940 Chem class : 8 Pack group : II
Quantity | 100mL |
---|
Chemical Identifiers
68-11-1 | |
92.112 | |
CWERGRDVMFNCDR-UHFFFAOYSA-N | |
1133 | |
2-sulfanylacetic acid |
C2H4O2S | |
4876 | |
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio | |
CHEBI:30065 | |
C(C(=O)O)S |
Specifications
Plastic Coated Glass bottle | |
Colorless | |
100mL | |
68-11-1 | |
4876 | |
CWERGRDVMFNCDR-UHFFFAOYSA-N | |
2-sulfanylacetic acid | |
1133 | |
92.11g/mol |
220°C | |
-16°C | |
1.5 | |
C2H4O2S | |
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio | |
C(C(=O)O)S | |
92.112 | |
CHEBI:30065 | |
Liquid |
Safety and Handling
- Mercaptoacetic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Special Offers
-
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.: 14802: 2021-02-28