missing translation for 'onlineSavingsMsg'
Learn More

Thioglycolic Acid, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7073608 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
This item is not returnable. View return policy

Product Code. 10020500

Brand: Fisher Chemical T/1000/PB05

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thioglycolic Acid, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-11-1
Molecular Formula C2H4O2S
Molecular Weight (g/mol) 92.112
MDL Number 4876
InChI Key CWERGRDVMFNCDR-UHFFFAOYSA-N
Synonym mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio
PubChem CID 1133
ChEBI CHEBI:30065
IUPAC Name 2-sulfanylacetic acid
SMILES C(C(=O)O)S

Specifications

Melting Point -16°C
Color Colorless
pH 1.5
Boiling Point 220°C
Packaging Plastic Coated Glass bottle
Quantity 100 mL
Formula Weight 92.11g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Mercaptoacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.