missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Thioflavin T

Product Code. 16441168
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441168 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441168 Supplier MedChemExpress Supplier No. HYD0218
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Thioflavin T is a cationic Benzothiazole dye that shows enhanced fluorescence upon binding to amyloid in tissue sections.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Product Type Dye Reagents
Quantity 5 g
Shipping Condition Room temperature
CAS 2390-54-7
Molecular Weight (g/mol) 318.86 g/mol
Synonym Basic Yellow 1
Target HBV
Solubility DMSO : 22.5 mg/mL (70.56 mM; Need ultrasonic) ∣ H2O : 12.5 mg/mL (39.20 mM; Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.