missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Thioflavin T

Product Code. 18738886
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18738886 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18738886 Supplier Tocris Bioscience™ Supplier No. 7122/500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fluorescent amyloid stain, binds to the stacked β sheets of amyloid fibrils

Thioflavin T is a cell-permeable fluorescent amyloid stain. This cationic benzothiazole dye increases in fluorescence upon binding to the stacked β sheets of amyloid fibrils. Thioflavin T is used to confirm formation of β sheet structure from mutant huntingtin exon-1 aggregates (Ex1Q48) in vitro. Thioflavin T is also used to monitor the kinetics of in vitro polyglutamine amyloid formation of tNhtt-42Q aggregates in a Huntington's disease cell model. Insoluble senile plaques of Aβ in brain tissues are commonly stained with Thioflavin T. Excitation/emission maxima = 440/485 nm.

This product is supplied with a high degree of hydration and some residual inorganic salts, the amount of which are batch dependent. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[4-(Dimethylamino)phenyl]-3,6-dimethylbenzothiazolium chloride
CAS 2390-54-7
Quantity 500 mg
Target Fluorescent Amyloid B Probes
Molecular Formula C17H19ClN2S
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.