missing translation for 'onlineSavingsMsg'
Learn More

Thiodiglycolic acid, 98%

Product Code. 10071640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071640 1 kg Plastic bottle 1kg
10071650 25 g Plastic bottle 25g
10326450 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10071640 Supplier Thermo Scientific Acros Supplier No. 138740010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-93-3 | C4H6O4S | 150.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 123-93-3
Molecular Formula C4H6O4S
Molecular Weight (g/mol) 150.148
MDL Number MFCD00004359
InChI Key UVZICZIVKIMRNE-UHFFFAOYSA-N
Synonym thiodiglycolic acid, 2,2'-thiodiacetic acid, thiodiacetic acid, mercaptodiacetic acid, tdga, dicarboxymethyl sulfide, thiodi acetic acid, carboxymethylthio acetic acid, 2,2'-thiodiethanoic acid, thiodiglycollic acid
PubChem CID 31277
ChEBI CHEBI:35012
IUPAC Name 2-(carboxymethylsulfanyl)acetic acid
SMILES C(C(=O)O)SCC(=O)O

Especificaciones

Melting Point 128.0°C to 131.0°C
Color Gray to White
Assay Percent Range 98%
Linear Formula S(CH2CO2H)2
Beilstein 03, 253
Merck Index 15, 9484
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in benzene
Formula Weight 150.15
Percent Purity 98%
Quantity 1 kg
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Chemical Name or Material Thiodiglycolic acid, 98%
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.