missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiocolchicoside, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Thiocolchicoside
Specifications
Specifications
| Chemical Name or Material | Thiocolchicoside |
| CAS | 602-41-5 |
| Analyte or Component Names | Thiocolchicoside |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H33 N O10 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | Colchicoside, 10-thio- (8CI), Colchicoside, thio- (6CI,7CI), Benzo[a]heptalene, acetamide deriv., 2-Demethoxy-2-glucosidoxythiocolchicine, Coltramyl, Coltrax, Etoshine MR, Miorel, Miotens, Muscoril, NSC 147755, R 271, Thiocholshikoside, Thiocolchicoside, Acetamide, N-[(7S)-3-(beta-D-glucopyranosyloxy)-5,6,7,9-tetrahydro-1,2-dimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]-, Acetamide, N-[3-(beta-D-glucopyranosyloxy)-5,6,7,9-tetrahydro-1,2-dimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]-, (S)- |
| InChI Formula | InChI=1S/C27H33NO10S/c1-12(30)28-16-7-5-13-9-18(37-27-24(34)23(33)22(32)19(11-29)38-27)25(35-2)26(36-3)21(13)14-6-8-20(39-4)17(31)10-15(14)16/h6,8-10,16,19,22-24,27,29,32-34H,5,7,11H2,1-4H3,(H,28,30)/t16-,19+,22+,23-,24+,27+/m0/s1 |
| SMILES | COc1c(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)cc3CC[C@H](NC(=O)C)C4=CC(=O)C(=CC=C4c3c1OC)SC |
| IUPAC Name | N-[(7S)-1,2-dimethoxy-10-methylsulfanyl-9-oxo-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction