missing translation for 'onlineSavingsMsg'
Learn More

Thiochroman-4-one, 98%

Product Code. 10240743 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10240743 10 g Glass bottle 10g
10619464 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10240743 Supplier Thermo Scientific Acros Supplier No. 153480100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3528-17-4 | C9H8OS | 164.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3528-17-4
Molecular Formula C9H8OS
Molecular Weight (g/mol) 164.22
MDL Number MFCD00006882
InChI Key CVQSWZMJOGOPAV-UHFFFAOYSA-N
Synonym thiochroman-4-one, 4h-1-benzothiopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-thiochromen-4-one, 3,4-dihydro-2h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one,3-dihydro, 2,3-dihydro-1-benzothiopyran-4-one, 4-thiochromanone, thiochroman-4-on, 1-thiochroman-4-one
PubChem CID 19048
IUPAC Name 2,3-dihydrothiochromen-4-one
SMILES C1CSC2=CC=CC=C2C1=O

Specifications

Melting Point 28°C to 30°C
Color Orange
Boiling Point 154°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6395
Quantity 10 g
Beilstein 17, II, 336
Formula Weight 164.22
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material Thiochroman-4-one
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.