missing translation for 'onlineSavingsMsg'
Learn More

Thiocarbohydrazide, 98%

Product Code. 10637304 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10637304 5 g Glass bottle 5g
10174640 25 g Plastic bottle 25g
10036293 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10637304 Supplier Thermo Scientific Acros Supplier No. 207530050
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2231-57-4 | CH6N4S | 106.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2231-57-4
Molecular Formula CH6N4S
Molecular Weight (g/mol) 106.15
MDL Number MFCD00007616
InChI Key LJTFFORYSFGNCT-UHFFFAOYSA-N
Synonym thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu
PubChem CID 2724189
ChEBI CHEBI:9549
IUPAC Name 1,3-diaminothiourea
SMILES NNC(=S)NN

Specifications

Melting Point 168.0°C to 172.0°C
Color Beige-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Glass bottle
Linear Formula H2NNHCSNHNH2
Quantity 5 g
Beilstein 03, 197
Formula Weight 106.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Thiocarbohydrazide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Thiocarbohydrazide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.