missing translation for 'onlineSavingsMsg'
Learn More

Thiocarbohydrazide, 97%

Product Code. 11374504
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374504 5 g 5g
11384504 25 g 25g
11394504 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11374504 Supplier Thermo Scientific Alfa Aesar Supplier No. L01205.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2231-57-4 | CH6N4S | 106.15 g/mol

Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiocarbohydrazide is used in electron microscopy to produce electron-opaque deposits for ultra structural analysis. It is used in the synthesis of oxazine grass ketones.

Solubility
Very slightly soluble (0.5g/100g, 25°C). Insoluble in ether.

Notes
Light Sensitive. Incompatible with strong acids, Strong oxidizing agents. Avoid heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2231-57-4
Molecular Formula CH6N4S
Molecular Weight (g/mol) 106.15
MDL Number MFCD00007616
InChI Key LJTFFORYSFGNCT-UHFFFAOYSA-N
Synonym thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu
PubChem CID 2724189
ChEBI CHEBI:9549
IUPAC Name 1,3-diaminothiourea
SMILES NNC(=S)NN

Specifications

Melting Point ∽175°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 506657
Sensitivity Light sensitive
Solubility Information Very slightly soluble (0.5g/100g,25°C). Insoluble in ether.
Formula Weight 106.15
Percent Purity 97%
Chemical Name or Material Thiocarbohydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.