missing translation for 'onlineSavingsMsg'
Learn More

Thiobencarb-d10 (diethyl-d10), TRC

Product Code. 30449282
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30449282

Brand: Toronto Research Chemicals TRCT34425610MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Thiobencarb diethyl-d10
CAS 1219804-12-2
Molecular Formula C12 D10 H6 Cl N O S
Quantity 10 mg
Synonym Carbamothioic acid','N,N-di(ethyl-1,1,2,2,2-d5)-','S-[(4-chlorophenyl)methyl] ester (ACI), S-[(4-Chlorophenyl)methyl] N,N-di(ethyl-1,1,2,2,2-d5)carbamothioate (ACI), Thiobencarb-d10, N,N-Di(ethyl-1,1,2,2,2-d5)-S-[(4-Chlorophenyl)methyl] Ester Carbamothioic Acid
InChI Formula InChI=1S/C12H16ClNOS/c1-3-14(4-2)12(15)16-9-10-5-7-11(13)8-6-10/h5-8H,3-4,9H2,1-2H3/i1D3,2D3,3D2,4D2
SMILES [2H]C([2H])([2H])C([2H])([2H])N(C(=O)SCc1ccc(Cl)cc1)C([2H])([2H])C([2H])([2H])[2H]
IUPAC Name S-[(4-chlorophenyl)methyl] N,N-bis(1,1,2,2,2-pentadeuterioethyl)carbamothioate
Molecular Weight (g/mol) 267.84
Formula Weight 267.1269
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.