missing translation for 'onlineSavingsMsg'
Learn More

Thioanisole, 99%

Product Code. 11473930
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2500 g
Unit Size:
100g
2500g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473930 25 g 25g
11492617 100 g 100g
11402627 500 g 500g
11412627 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11473930 Supplier Thermo Scientific Alfa Aesar Supplier No. A14846.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 100-68-5 | C7H8S | 124.201 g/mol

Thioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.

Solubility
Miscible with most common organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-68-5
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.201
MDL Number MFCD00008559
InChI Key HNKJADCVZUBCPG-UHFFFAOYSA-N
Synonym thioanisole, methyl phenyl sulfide, methylthio benzene, thioanisol, benzene, methylthio, phenylthiomethane, phenyl methyl sulfide, methylsulfanyl benzene, methylphenylsulfide, 1-thiaethyl benzene
PubChem CID 7520
IUPAC Name methylsulfanylbenzene
SMILES CSC1=CC=CC=C1

Specifications

Melting Point -15°C
Density 1.058
Boiling Point 188°C to 189°C
Flash Point 73°C (163°F)
Odor Stench
Refractive Index 1.586
Quantity 25 g
UN Number UN1993
Beilstein 1904179
Solubility Information Miscible with most common organic solvents. Immiscible with water.
Formula Weight 124.21
Percent Purity 99%
Chemical Name or Material Thioanisole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thioanisole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Thioanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.