missing translation for 'onlineSavingsMsg'
Learn More

Thioanisole, 99%

Product Code. 11402627
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2500 g
Unit Size:
100g
2500g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11402627 500 g 500g
11473930 25 g 25g
11492617 100 g 100g
11412627 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11402627 Supplier Thermo Scientific Alfa Aesar Supplier No. A14846.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 100-68-5 | C7H8S | 124.201 g/mol

Thioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioanisole is used as an intermediate in the preparation of dyes, pharmaceuticals and agrochemicals. It is also used to prepare stable sulfonium salts, sulfoxides and sufones. It is utilized for the cleavage of methyl ethers in association with triflic acid. It acts as a flavoring agent or an adjuvant. It is an important starting material for the synthesis of 3-substituted benzo[b]thiophenes.

Solubility
Miscible with most common organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-68-5
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.201
MDL Number MFCD00008559
InChI Key HNKJADCVZUBCPG-UHFFFAOYSA-N
Synonym thioanisole, methyl phenyl sulfide, methylthio benzene, thioanisol, benzene, methylthio, phenylthiomethane, phenyl methyl sulfide, methylsulfanyl benzene, methylphenylsulfide, 1-thiaethyl benzene
PubChem CID 7520
IUPAC Name methylsulfanylbenzene
SMILES CSC1=CC=CC=C1

Specifications

Melting Point -15°C
Density 1.058
Boiling Point 188°C to 189°C
Flash Point 73°C (163°F)
Odor Stench
Refractive Index 1.586
Quantity 500 g
UN Number UN1993
Beilstein 1904179
Solubility Information Miscible with most common organic solvents. Immiscible with water.
Formula Weight 124.21
Percent Purity 99%
Chemical Name or Material Thioanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.