missing translation for 'onlineSavingsMsg'
Learn More

Thioacetamide, 98%

Product Code. 11429753
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429753 25 g 25g
11439753 100 g 100g
11449753 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429753 Supplier Thermo Scientific Alfa Aesar Supplier No. A12926.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 62-55-5 | C2H5NS | 75.13 g/mol

Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.

Solubility
Soluble in ethanol, benzene and petroleum ether. Slightly soluble in water and ether.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-55-5
Molecular Formula C2H5NS
Molecular Weight (g/mol) 75.13
MDL Number MFCD00008070
InChI Key YUKQRDCYNOVPGJ-UHFFFAOYSA-N
Synonym thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
PubChem CID 2723949
ChEBI CHEBI:32497
IUPAC Name ethanethioamide
SMILES CC(N)=S

Specifications

Melting Point 110°C to 114°C
Density 1.37
Assay Percent Range 98%
Quantity 25 g
Beilstein 506006
Merck Index 14,9319
Solubility Information Soluble in ethanol,benzene and petroleum ether; Slightly soluble in water and ether.
Formula Weight 75.13
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Thioacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thioacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Thioacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.