missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Thinline Slotted Cassettes in Tube Packs

Product Code. 16394106 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
16394106 Yellow Pack of 750
16384096 Beige Pack of 750
16394096 Blue Pack of 750
16304106 Green Pack of 750
16314106 Gray Pack of 750
16324106 Lavender Pack of 750
16334106 Light Green Pack of 750
16344106 Lilac Pack of 750
16354106 Orange Pack of 750
16364106 Pink Pack of 750
16374106 Tan Pack of 750
16384106 White Pack of 750
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 16394106 Supplier Epredia™ Supplier No. B851736YW
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For routine tissue samples, cassettes are compatible with all Epredia PrintMate™ Cassette Printers and have a front lid hinge with color-matched, attached lids available in 12 different color varieties.

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY

Specifications

Pore Style Slots
Closure Included Yes
Quantity 750/Cs.
Closure Type Attached Lid, Front Hinge
PrintMate Compatible Yes
Color Yellow
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Slotted Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.