missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Thinline Biopsy Cassettes in Tube Packs

Product Code. 16304126 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Light Green
Lilac
Orange
Pink
Tan
White
Unit Size:
Pack of 750
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
16304126 White Pack of 750
16304116 Beige Pack of 750
16314116 Blue Pack of 750
16324116 Green Pack of 750
16334116 Gray Pack of 750
16354116 Light Green Pack of 750
16364116 Lilac Pack of 750
16374116 Orange Pack of 750
16384116 Pink Pack of 750
16394116 Tan Pack of 750
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 16304126 Supplier Epredia™ Supplier No. B851737WH

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For biopsy tissue samples, cassettes have a front lid hinge, attached lids in matching colors, and are compatible with all Epredia PrintMate™ Cassette Printers

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY

Specifications

Pore Style Small Squares
Closure Included Yes
Quantity 750/Cs.
Closure Type Attached Lid, Front Hinge
PrintMate Compatible Yes
Color White
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Thinline Biopsy Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.