Learn More
Greiner Bio-One ThinCert™ Tissue Culture Inserts
Product Code. p-7075424
Click to view available options
Pore Size:
0.4 μm
1.0 μm
3 μm
3.0 μm
8.0 μm
No. of Wells:
6
12
24
Pore Density:
0.15 x 106 pores/cm2
0.6 x 106 pores/cm2
1 x 108 pores/cm2
2 x 106 pores/cm2
Unit Size:
Pack of 24
Pack of 48
Product Code. 10689094
Brand: Greiner Bio-One 657630
Only null left
Add to basket
Add to basket
Research tool for transport, co-culture, migration and invasion studies. Greiner Bio-One ThinCert™ Tissue Culture Inserts provides improved cell adhesion due to physical surface treatment.
  • Clear polystyrene housing with sealed PET capillary pore membrane
  • Hanging geometry provides protection to lower compartment cell layer
  • Simplified pipetting due to self-lift geometry
  • Eccentric in-well positioning allows unequalled access to lower compartment
  • Available in 6, 12 and 24 well format
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene housing with sealed PET capillary pore membrane
Pore Density 0.6 x 106 pores/cm2
Pore Size 3 μm
No. of Wells 6