missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One ThinCert™ 96 Well HTS Insert

Product Code. 18808366
Change view
Click to view available options
Product Type:
Insert with Permeable Membrane
Surface Treatment:
Tissue Culture
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Surface Treatment
18808366 Insert with Permeable Membrane Tissue Culture
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18808366 Supplier Greiner Bio-One Supplier No. 655640

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thincert™ 96 Well HTS inserts are indispensable tools for high-throughput screening and assays. Designed for precision, they enable researchers to elevate the quality and efficiency of their work.

  • Optimal tool for generating tissue models (e.g., endothelia and epithelia) for transport, uptake, or co-culture studies in submersed as well as air-lift culture with an automation-friendly format
  • By combining high membrane permeability and best possible transparency, Greiner Bio-One is offering a unique 96 well insert version, exclusively designed for applications that require both high diffusion rates as well as microscopic readouts.
  • Special pore arrangement and a high pore density of 2 x 107/cm2.
  • Patented assembly technology: robust and reliable bonding of membrane 
  • Excellent membrane flatness: optimal and reproducible cell culture conditions 
  • Optimized fit: prevention of media wicking
  • Enlarged access ports: convenient access for pipettes, robots and TEER electrodes
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Color Clear
Product Type Insert with Permeable Membrane
Material Polycarbonate
Volume (Metric) Well 0.15 to 1.7 μL
No. of Wells 96
Volume (Metric) Working 15 to 160 μL
Sterility Sterile
Surface Treatment Tissue Culture
Well Shape Flat Bottom (F-well)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.