missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thimet Oligopeptidase/THOP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | Thimet Oligopeptidase/THOP1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 4-6 μL/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18644992
|
Novus Biologicals
NBP2-98355-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640415
|
Novus Biologicals
NBP2-98355-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thimet Oligopeptidase/THOP1 Polyclonal antibody specifically detects Thimet Oligopeptidase/THOP1 in Mouse samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| Thimet Oligopeptidase/THOP1 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 7064 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 4-6 μL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| EC 3.4.24, EC 3.4.24.15, Endopeptidase 24.15, EP24.15, MEPD_HUMAN, MP78, thimet oligopeptidase, thimet oligopeptidase 1, TOP | |
| Produced in rabbits immunized with purified, recombinant Mouse Thimet Oligopeptidase/THOP1 (Accession#: NP_073144.3; Lys 2-Cys687) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title