missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thimet Oligopeptidase/THOP1 Antibody (4D6), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB400-146AF405
This item is not returnable.
View return policy
Description
Thimet Oligopeptidase/THOP1 Monoclonal specifically detects Thimet Oligopeptidase/THOP1 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Thimet Oligopeptidase/THOP1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| P52888 | |
| THOP1 | |
| A synthetic peptide corresponding to residues 56-72 of human Thimet Oligopeptidase. [UniProt# P52888] | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate, Rabbit | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 4D6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| EC 3.4.24, EC 3.4.24.15, Endopeptidase 24.15, EP24.15, MEPD_HUMAN, MP78, thimet oligopeptidase, thimet oligopeptidase 1, TOP | |
| Mouse | |
| 80 kDa | |
| 0.1 mL | |
| Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 7064 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction