missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Thimet Oligopeptidase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583269
This item is not returnable.
View return policy
Description
Immunogen sequence: AACAGDMADA ASPCSVVNDL RWDLSAQQIE ERTRELIEQT KRVYDQVGTQ EFEDVSYEST LKA Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 76%.
A steady-state level of amyloid beta-peptide (Abeta) represents a balance between its biosynthesis from the amyloid precursor protein (APP) through the action of the beta and gamma-secretases and its catabolism by a variety of proteolytic enzymes. Thimet Oligopeptidase (THOP1) has been shown to promote A-beta degradation. It has also been shown to play a role in antigen presentation, metabolism of neuropeptides under 20 amino acids, and cytoplasmic peptide degradation.
Specifications
| Thimet Oligopeptidase | |
| Polyclonal | |
| Unconjugated | |
| Thop1 | |
| AI131655; AI327041; EC 3.4.24; EC 3.4.24.15; endo-oligopeptidase A; Endopeptidase 24.15; EP24.15; MEPD_HUMAN; metalloendopeptidase; MP78; PZ-peptidase; soluble metallo-endopeptidase; Thimet oligopeptidase; thimet oligopeptidase 1; Thop1; TOP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7064 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P52888 | |
| Thop1 | |
| Recombinant protein corresponding to Human THOP1. Recombinant protein control fragment (Product #RP-104658). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction