missing translation for 'onlineSavingsMsg'
Learn More

Thimerosal

Product Code. p-7042232
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 15414319

Brand: Thermo Scientific Alfa Aesar J61799.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thimerosal, CAS # 54-64-8, is a sulfhydryl (thiol) oxidizing reagent with antibacterial and antifungal activities that is used to protect components from chemical change or microbial action. | CAS: 54-64-8 | C9H9HgNaO2S | 404.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• Thimerosal is an organic compound and a derivative of thiosalicyclic acid. It shows antibacterial and antifungal activities and has the ability to protect against chemical change or microbial action

• Thimerosal can inhibit the sulfhydryl-containing active site of some enzymes and bind to sulfhydryl compounds, such as glutathione, cysteine, and thiol groups of proteins

• It can activate the InsP3 calcium channel on the endoplasmic reticular membrane, triggering the release of calcium from intracellular stores, leading to a calcium-influx of extracellular calcium. Thus, thimerosal may stimulate or inhibit cellular functions dependent on calcium signaling

Application

• Thimerosal can increase histamine release and cell-mediated immunity

• It has the ability to inhibit various sulfhydryl-dependent enzymes and proteins, including PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na+/K+-dependent ATPases

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-64-8
Molecular Formula C9H9HgNaO2S
Molecular Weight (g/mol) 404.81
MDL Number MFCD00013062
InChI Key RTKIYNMVFMVABJ-UHFFFAOYSA-L
Synonym thimerosal, thiomersal, thiomersalate, mercurothiolate, sodium merthiolate, sodium ethylmercurithiosalicylate, merthiolate, thimerosalate, thimerosalum, thimersalate
PubChem CID 16684434
ChEBI CHEBI:9546
SMILES [Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O

Specifications

Melting Point 234°C to 237°C
Color White
Flash Point 250°C (482°F)
Quantity 10 g
UN Number UN2025
Beilstein 8169555
Merck Index 14,9317
Solubility Information Soluble in water or ethanol
IUPAC Name sodium [(2-carboxylatophenyl)sulfanyl](ethyl)mercury
Formula Weight 404.81
Physical Form Powder
Chemical Name or Material Thimerosal
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Thimerosal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sodium o-(ethylmercurithio)benzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.