missing translation for 'onlineSavingsMsg'
Learn More

Thiethylperazine Dimaleate, TRC

Product Code. 30453085
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453085 100 mg -
30452247 25 mg 25mg
30455791 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453085 Supplier Toronto Research Chemicals Supplier No. TRCT344505100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1179-69-7
Molecular Formula C22 H29 N3 S2 . 2 C4 H4 O4
Molecular Weight (g/mol) 631.76
Synonym 10H-Phenothiazine, 2-(ethylthio)-10-[3-(4-methyl-1-piperazinyl)propyl]-, (Z)-2-butenedioate (1:2), Phenothiazine, 2-(ethylthio)-10-[3-(4-methyl-1-piperazinyl)propyl]-, dimaleate (6CI), Phenothiazine, 2-(ethylthio)-10-[3-(4-methyl-1-piperazinyl)propyl]-, maleate (1:2) (7CI,8CI), 2-(Ethylthio)-10-[3-(4-methyl-1-piperazinyl)propyl]phenothiazine dimaleate, 2-Ethylmercapto-10-[3-(1-methyl-4-piperazinyl)propyl]phenothiazine dimaleate, GS 95, NSC 130044, Thiethylperazine dimaleate, Thiethylperazine maleate, Torecan, Torecan maleate, Toresten, Tresten
IUPAC Name (Z)-but-2-enedioic acid;2-ethylsulfanyl-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine
SMILES CCSc1ccc2Sc3ccccc3N(CCCN4CCN(C)CC4)c2c1.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H29N3S2.2C4H4O4/c1-3-26-18-9-10-22-20(17-18)25(19-7-4-5-8-21(19)27-22)12-6-11-24-15-13-23(2)14-16-24;2*5-3(6)1-2-4(7)8/h4-5,7-10,17H,3,6,11-16H2,1-2H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-
Formula Weight 631.2022
Chemical Name or Material Thiethylperazine Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.