missing translation for 'onlineSavingsMsg'
Learn More

Thietan-3-ol, TRC

Product Code. 30455295
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
50mg
Product Code. Quantity unitSize
30455295 500 mg -
30454349 50 mg 50mg
30453423 100 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 30455295

Brand: Toronto Research Chemicals TRCT344488500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10304-16-2
Formula Weight 90.0139
InChI Formula InChI=1S/C3H6OS/c4-3-1-5-2-3/h3-4H,1-2H2
IUPAC Name thietan-3-ol
Molecular Formula C3H6OS
Molecular Weight (g/mol) 90.14
SMILES OC1CSC1
Chemical Name or Material Thietan-3-ol

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.