missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thieno[2,3-b]pyrazine-6-carboxylic acid, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 180.19 |
Percent Purity | 90% |
Chemical Name or Material | Thieno[2, 3-b]pyrazine-6-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'