missing translation for 'onlineSavingsMsg'
Learn More

Thidiazuron, TRC

Product Code. 17983905
Change view
Click to view available options
Quantity:
100 mg
500 mg
5 g
Unit Size:
100mg
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983905 100 mg 100mg
17952855 5 g 5g
17901885 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17983905 Supplier Toronto Research Chemicals Supplier No. TRCT344275100MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51707-55-2
Molecular Formula C9 H8 N4 O S
Molecular Weight (g/mol) 220.25
Synonym 1,2,3-Thiadiazole, urea deriv., 1-Phenyl-3-(1,2,3-thiadiazol-5-yl)urea, Avguron, CCG-24904, Dropp, Dropp SC, Gross, Lift, N-Phenyl-N'-1,2,3-thiadiazol-5-ylurea, SN 49537, TAG, TAG (plant growth regulator), TDZ, Thidiazuron, Urea, N-phenyl-N'-1,2,3-thiadiazol-5-yl-
IUPAC Name 1-phenyl-3-(thiadiazol-5-yl)urea
SMILES O=C(Nc1ccccc1)Nc2cnns2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H8N4OS/c14-9(12-8-6-10-13-15-8)11-7-4-2-1-3-5-7/h1-6H,(H2,11,12,14)
Formula Weight 220.0419
Percent Purity >95%
Chemical Name or Material Thidiazuron
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.