missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Thiazovivin

Product Code. 17129971
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 17129971

Brand: STEMCELL Technologies 1000247

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RHO/ROCK pathway inhibitor; Inhibits ROCK

  • Thiazovivin is a selective inhibitor of Rho-associated coiled-coil containing protein kinase (ROCK), a serine/threonine kinase that plays a role in cell polarity, contraction, and actin cytoskeleton reorganization (Xu et al.) Thiazovivin is effective at 5-fold-lower concentrations than another common ROCK inhibitor Y-27632 (Catalog #72302; Xu et al.)
  • Promotes survival of human embryonic stem (ES) cells during dissociation by stabilizing E-cadherin and improves cell attachment (Xu et al.)
  • Promotes survival of single human induced pluripotent stem (iPS) cells during transfection for TALEN-mediated genome editing (Sun and Zhao)
  • Increases the efficiency of reprogramming human somatic cells to iPS cells, in combination with PD0325091 and SB431542 (Lin et al.)
  • Increases the efficiency of reprogramming human cord blood mononuclear cells to iPS cells (Hu et al.)

Specifications

CAS 1226056-71-8
Synonym Tzv
Quantity 25 mg
Molecular Formula C15H13N5OS
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.