missing translation for 'onlineSavingsMsg'
Learn More

Thiazolidine, TRC

Product Code. 30133819
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133819 50 g 50g
30137204 25 g 25g
30005322 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133819 Supplier Toronto Research Chemicals Supplier No. TRCT34424550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-78-9
Molecular Formula C3H7NS
Molecular Weight (g/mol) 89.16
Synonym 1,3-Thiazolidine, 1-Thia-3-azacyclopentane, Tetrahydrothiazole, Tetrahydro-thiazole
IUPAC Name 1,3-thiazolidine
SMILES S1CNCC1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C3H7NS/c1-2-5-3-4-1/h4H,1-3H2
Formula Weight 89.0299
Chemical Name or Material Thiazolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.