missing translation for 'onlineSavingsMsg'
Learn More

Thiazolidine-2-carboxylic acid, 97%

Product Code. 11494613
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494613 100 g 100g
11474613 5 g 5g
11484613 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494613 Supplier Thermo Scientific Alfa Aesar Supplier No. A11564.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16310-13-7 | C4H7NO2S | 133.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16310-13-7
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.165
MDL Number MFCD00068005
InChI Key ULSZVNJBVJWEJE-UHFFFAOYSA-N
Synonym thiazolidine-2-carboxylic acid, 1,3-thiazolane-2-carboxylic acid, 2-thiazolidinecarboxylic acid, beta-thiaproline, thiazolidine-2-carboxylicacid, acmc-20aiwa, thiazolidine carboxylic acid, 2-triazolidinecarboxylic acid, ulszvnjbvjweje-uhfffaoysa, tetrahydrothiazole-2-carboxylic acid
PubChem CID 42486
IUPAC Name 1,3-thiazolidine-2-carboxylic acid
SMILES C1CSC(N1)C(=O)O

Specifications

Melting Point ∼184°C (decomposition)
Quantity 100 g
Beilstein 112783
Formula Weight 133.17
Percent Purity 97%
Chemical Name or Material Thiazolidine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.