missing translation for 'onlineSavingsMsg'
Learn More

Thiazole, 99%

Product Code. 10679853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10679853 25 g Glass bottle 25g
10438730 1 g Glass bottle 1g
10021890 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10679853 Supplier Thermo Scientific Acros Supplier No. 166130250
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 288-47-1 | C3H3NS | 85.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-47-1
Molecular Formula C3H3NS
Molecular Weight (g/mol) 85.12
MDL Number MFCD00005315
InChI Key FZWLAAWBMGSTSO-UHFFFAOYSA-N
Synonym thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium
PubChem CID 9256
ChEBI CHEBI:43732
IUPAC Name 1,3-thiazole
SMILES S1C=CN=C1

Specifications

Color Colorless to Yellow
Density 1.1980g/mL
Boiling Point 117.0°C to 118.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5365 to 1.5395
Quantity 25 g
Beilstein 27, 15
Merck Index 15, 9459
Specific Gravity 1.198
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 85.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Thiazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Thiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.