missing translation for 'onlineSavingsMsg'
Learn More

Thiazole, 99%

Product Code. 11332377
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332377 5 g 5g
11322377 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332377 Supplier Thermo Scientific Alfa Aesar Supplier No. L09970.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 288-47-1 | C3H3NS | 85.12 g/mol

Thiazole is used as a flavoring agent and in the preparation of dyes and rubber accelerators. It serves as a component of the vitamin thiamine (B1). It acts as a protected formyl group used in natural product synthesis. It reacts with alkyl lithium and Grignard’s reagent to prepare organometallic complexes. It is involved in the electrophilic aromatic substitution and nucleophilic aromatic substitution at C-5 and C-2 positions respectively. Further, it undergoes alkylation reaction to get thiazolium cation, which is used as a catalyst in the Stetter reaction and the Benzoin condensation. In addition to this, it is involved in the preparation of alagebrium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole is used as a flavoring agent and in the preparation of dyes and rubber accelerators. It serves as a component of the vitamin thiamine (B1). It acts as a protected formyl group used in natural product synthesis. It reacts with alkyl lithium and Grignard’s reagent to prepare organometallic complexes. It is involved in the electrophilic aromatic substitution and nucleophilic aromatic substitution at C-5 and C-2 positions respectively. Further, it undergoes alkylation reaction to get thiazolium cation, which is used as a catalyst in the Stetter reaction and the Benzoin condensation. In addition to this, it is involved in the preparation of alagebrium.

Solubility
Soluble in ethanol and ether. Slightly soluble in water.

Notes
Air and light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-47-1
Molecular Formula C3H3NS
Molecular Weight (g/mol) 85.12
MDL Number MFCD00005315
InChI Key FZWLAAWBMGSTSO-UHFFFAOYSA-N
Synonym thiazole, thiazol, unii-320rcw8pef, fema no. 3615, ccris 3205, 320rcw8pef, lom, racemic thiazole, 5h-thiazol-1-ium
PubChem CID 9256
ChEBI CHEBI:43732
IUPAC Name 1,3-thiazole
SMILES S1C=CN=C1

Specifications

Melting Point -33°C
Density 1.2
Boiling Point 117°C to 118°C
Flash Point 22°C (71°F)
Odor Unpleasant
Refractive Index 1.539
Quantity 5 g
UN Number UN1993
Beilstein 103852
Sensitivity Air and light sensitive
Merck Index 14,9307
Solubility Information Soluble in ethanol and ether. Slightly soluble in water.
Formula Weight 85.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Thiazole
Show More Show Less

RUO – Research Use Only

Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.