missing translation for 'onlineSavingsMsg'
Learn More

Thiazole-5-methanol, 95%, Thermo Scientific™

Product Code. 15335018 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15335018 5 g 5g
15345018 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15335018 Supplier Thermo Scientific Acros Supplier No. 455550050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 38585-74-9
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
InChI Key WKBQQWDVVHGWDB-UHFFFAOYSA-N
Synonym 5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl-1,3-thiazole
PubChem CID 2763216
IUPAC Name 1,3-thiazol-5-ylmethanol
SMILES C1=C(SC=N1)CO

Specifications

Density 1.3200g/mL
Assay Percent Range 94% min. (GC)
Quantity 5 g
Specific Gravity 1.32
Formula Weight 115.15
Percent Purity 95%
Chemical Name or Material Thiazole-5-methanol
compliance-icons
Product Identifier
  • Thiazole-5-methanol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.