missing translation for 'onlineSavingsMsg'
Learn More

Thiazole-2-carboxaldehyde, 95%, Thermo Scientific Chemicals

Product Code. 15489968
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Thiazole-2-carboxaldehyde may be used as a reactant in the following syntheses Benzothiazine N-acylhydrazones, having potential antinociceptive and anti-inflammatory activity, Thiazole-2-yl-(amino)methylphosphonate diethyl esters, Imino ester by reacting with L-leucine t-butyl ester hydrochloride. Also used in building block for taxane analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole-2-carboxaldehyde may be used as a reactant in the following syntheses Benzothiazine N-acylhydrazones, having potential antinociceptive and anti-inflammatory activity, Thiazole-2-yl-(amino)methylphosphonate diethyl esters, Imino ester by reacting with L-leucine t-butyl ester hydrochloride. Also used in building block for taxane analogs.

Solubility
Fully miscible with water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10200-59-6
Molecular Formula C4H3NOS
Molecular Weight (g/mol) 113.134
MDL Number MFCD00142924
InChI Key ZGTFNNUASMWGTM-UHFFFAOYSA-N
Synonym 2-thiazolecarboxaldehyde, thiazole-2-carbaldehyde, 2-formylthiazole, thiazole-2-carboxaldehyde, 2-formyl thiazole, thiazolecarboxaldehyde, 2-formyl-1,3-thiazole, 1,3-thiazole-2-carboxaldehyde, 2-thiazolecarbaldehyde, pubchem9929
PubChem CID 2734903
ChEBI CHEBI:43623
IUPAC Name 1,3-thiazole-2-carbaldehyde
SMILES C1=CSC(=N1)C=O

Specifications

Color Colorless to Yellow
Density 1.288 g/mL
Boiling Point 61°C to 63°C (15 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.574
Quantity 5 g
Solubility Information Fully miscible with water.
Formula Weight 113.13
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Thiazole-2-carboxaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'