missing translation for 'onlineSavingsMsg'
Learn More

Thiazol-5-ylmethanol, Mikromol™

Product Code. 30051550
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051550 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051550 Supplier Mikromol Supplier No. MM1034.220100
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Thiazol-5-ylmethanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Thiazolemethanol
CAS 38585-74-9
Analyte or Component Names 5-Thiazolemethanol
Shipping Condition Ice pack (-20°C)
Molecular Formula C4 H5 N O S
Certifications/Compliance ISO 9001
Synonym 5-Thiazolemethanol, (1,3-Thiazol-5-yl)methanol, 5-(Hydroxymethyl)-1,3-thiazole, 5-(Hydroxymethyl)thiazole, Thiazol-5-ylmethanol
InChI Formula InChI=1S/C4H5NOS/c6-2-4-1-5-3-7-4/h1,3,6H,2H2
SMILES OCc1cncs1
IUPAC Name 1,3-thiazol-5-ylmethanol
Molecular Weight (g/mol) 115.15
Quantity 100 mg
Formula Weight 115.0092 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.