missing translation for 'onlineSavingsMsg'
Learn More

Thianthrene-1-boronic acid, tech. 90%

Product Code. 11339815
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339815 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11339815 Supplier Thermo Scientific Alfa Aesar Supplier No. L19833.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 108847-76-3 | C12H9BO2S2 | 260.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108847-76-3
Molecular Formula C12H9BO2S2
Molecular Weight (g/mol) 260.132
MDL Number MFCD00093039
InChI Key FZEWPLIHPXGNTB-UHFFFAOYSA-N
Synonym thianthrene-1-boronic acid, 1-thianthrenylboronic acid, thianthren-1-boronic acid, boronic acid, 1-thianthrenyl, thianthren-1-yl boronic acid, pubchem11895, acmc-1c7cr, boronic acid, b-1-thianthrenyl
PubChem CID 2734382
IUPAC Name thianthren-1-ylboronic acid
SMILES B(C1=C2C(=CC=C1)SC3=CC=CC=C3S2)(O)O

Specifications

Melting Point 145°C to 149°C
Quantity 5 g
Beilstein 225524
Formula Weight 260.14
Percent Purity 90%
Grade Technical
Chemical Name or Material Thianthrene-1-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.