missing translation for 'onlineSavingsMsg'
Learn More

Thianaphthene, 97%

Product Code. 10173520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173520 5 g Glass bottle 5g
10636342 25 g Glass bottle 25g
10582154 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10173520 Supplier Thermo Scientific Acros Supplier No. 138620050
 Restricted Item
In Stock
Add to basket
Tilføj til indkøbskurv
Request Bulk or Custom Formats
Denne vare kan ikke returneres. Se returpolitik

CAS: 95-15-8 | C8H6S | 134.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 95-15-8
Molecular Formula C8H6S
Molecular Weight (g/mol) 134.2
MDL Number MFCD00005864
InChI Key FCEHBMOGCRZNNI-UHFFFAOYSA-N
Synonym benzo b thiophene, thianaphthene, benzothiophene, benzothiofuran, thionaphthene, 1-thiaindene, thianaphthen, benzothiophen, thianaphtene, 1-benzothiophen
PubChem CID 7221
ChEBI CHEBI:35858
IUPAC Name 1-benzothiophene
SMILES C1=CC=C2C(=C1)C=CS2

Tekniske data

Melting Point 28.0°C to 32.0°C
Color Pink to White or Yellow
Density 1.1400g/mL
Boiling Point 221.0°C to 222.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 17, 59
Merck Index 15, 9455
Specific Gravity 1.14
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in usual organic solvents, soluble in ethanol, ethylether, acetone
Formula Weight 134.2
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material Thianaphthene
Vis mere Vis mindre
compliance-icons compliance-icons
Produktidentifikator
  • Thianaphthene
Signalord
  • Warning
Farekategori
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Faresætning
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Sikkerhedssætning
  • P264-Wash face, hands and any thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to an approved waste d.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.