missing translation for 'onlineSavingsMsg'
Learn More

Thiamphenicol, Thermo Scientific™

The methyl-sulfonyl analogue of chloramphenicol

Brand:  Thermo Scientific Alfa Aesar J63575.03

Quantity 1g

 View more versions of this product

Product Code. 15485769

  • £23.90 / 1g
 Restricted Item
Estimated Shipment: 15-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Thiamphenicol is used to treat chancroid in men and uncomplicated gonorrhea. It is used in studies of bacterial protein synthesis at the level of peptidyl transferase activity associated with the 23S rRNA of the 50S ribosomal subunit. It is used to study chloraniphenicol-thiamphenicol-resistance and the use of fluorinated analogs when resistance is encountered. Its main advantage over chloramphenicol is that it has never been associated with aplastic anaemia.

Solubility
Soluble in acetonitrile or DMF. Slightly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

15318-45-3
356.214
OTVAEFIXJLOWRX-NXEZZACHSA-N
27200
2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
C12H15Cl2NO5S
MFCD00467983
thiamphenicol, thiophenicol, thiamcol, +-thiamphenicol, raceophenidol, thiocymetin, dextrosulphenidol, thiamphenicolum, d-thiocymetin, d-thiophenicol
CHEBI:32215
CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O

Specifications

15318-45-3
MFCD00467983
2819542
thiamphenicol, thiophenicol, thiamcol, +-thiamphenicol, raceophenidol, thiocymetin, dextrosulphenidol, thiamphenicolum, d-thiocymetin, d-thiophenicol
OTVAEFIXJLOWRX-NXEZZACHSA-N
2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
27200
356.22
C12H15Cl2NO5S
1g
14,9301
Soluble in acetonitrile or DMF; Slightly soluble in water
CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O
356.214
CHEBI:32215
Thiamphenicol
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.