missing translation for 'onlineSavingsMsg'
Learn More

Thiamphenicol, MedChemExpress

Product Code. 16428336
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428336 100 mg 100mg
16412156 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16428336 Supplier Medchem Express Supplier No. HYB0479
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Thiamphenicol (Thiophenicol), a methyl-sulfonyl derivative of Chloramphenicol, is a broad-spectrum antimicrobial antibiotic. Thiamphenicol acts by binding to the 50S ribosomal subunit, leading to inhibition of protein synthesis and bacteriostatic effect (against Gram-negative, Gram-positive aerobic and anaerobic bacteria).

Thiamphenicol (Thiophenicol), a methyl-sulfonyl derivative of Chloramphenicol, is a broad-spectrum antimicrobial antibiotic. Thiamphenicol acts by binding to the 50S ribosomal subunit, leading to inhibition of protein synthesis and bacteriostatic effect (against Gram-negative, Gram-positive aerobic and anaerobic bacteria).

Specifications

Chemical Name or Material Thiamphenicol
Physical Form Solid
CAS 15318-45-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H15Cl2NO5S
For Use With (Application) COVID-19-immunoregulation
Synonym Thiophenicol Dextrosulphenidol
Solubility Information DMSO : 100 mg/mL (280.73 mM; Need ultrasonic) ∣H2O : 2 mg/mL (5.61 mM; Need ultrasonic)
SMILES O=C(N[C@H](CO)[C@H](O)C1=CC=C(S(=O)(C)=O)C=C1)C(Cl)Cl
Molecular Weight (g/mol) 356.22
Formula Weight 356.22
Percent Purity 99.38%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.