missing translation for 'onlineSavingsMsg'
Learn More

Thiamphenicol, 99.5%, MP Biomedicals™

Product Code. p-4405260 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11465251

Brand: MP Biomedicals 0215684605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 15318-45-3
Molecular Formula C12H15Cl2NO5S
Molecular Weight (g/mol) 356.214
InChI Key OTVAEFIXJLOWRX-NXEZZACHSA-N
Synonym D -threo-2, 2-dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-(methyl- sulfonyl)phenethyl]-acetamide
PubChem CID 27200
ChEBI CHEBI:32215
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(C(CO)NC(=O)C(Cl)Cl)O

Specifications

Type Thiamphenicol
Physical Form Free-flowing Powder
Color White
Quantity 5 g
Assay Percent Range 0.995
Formula Weight 356.2
Chemical Name or Material Thiamphenicol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.