missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Nitrate, TRC

Product Code. 30006059
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006059 10 g 10g
17988394 5 g 5g
17918204 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006059 Supplier Toronto Research Chemicals Supplier No. TRCT34410510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-43-4
Molecular Formula C12 H17 N4 O S . N O3
Molecular Weight (g/mol) 327.36
Synonym Thiamine Nitrate, Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, nitrate (1:1), Thiamine nitrate (6CI), Thiamine nitrate (salt) (7CI,8CI), Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, nitrate (salt) (9CI), 3-(4-Amino-2-methylpyrimidyl-5-methyl)-4-methyl-5,β-hydroxyethylthiazolium nitrate, Aneurine mononitrate, Aneurine-nitrate, Betabion mononitrate, Thiamine mononitrate, Vitamin B1 mononitrate, Vitamin B1 nitrate
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;nitrate
SMILES Cc1ncc(C[n+]2csc(CCO)c2C)c(N)n1.[O-][N+](=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H17N4OS.NO3/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;2-1(3)4/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);/q+1;-1
Formula Weight 327.1001
Chemical Name or Material Thiamine Mononitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.