missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiamine Nitrate, Mikromol™
Description
Thiamine Nitrate
Specifications
Specifications
| Chemical Name or Material | Thiamine Mononitrate |
| CAS | 532-43-4 |
| Analyte or Component Names | Thiamine Mononitrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H17 N4 O S . N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Thiamine Nitrate, Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, nitrate (1:1), Thiamine nitrate (6CI), Thiamine nitrate (salt) (7CI,8CI), Thiazolium, 3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, nitrate (salt) (9CI), Aneurine mononitrate, Aneurine-nitrate, Betabion mononitrate, Thiamine mononitrate, Vitamin B1 mononitrate, Vitamin B1 nitrate, 3-(4-Amino-2-methylpyrimidyl-5-methyl)-4-methyl-5,beta-hydroxyethylthiazolium nitrate |
| InChI Formula | InChI=1S/C12H17N4OS.NO3/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;2-1(3)4/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);/q+1;-1 |
| SMILES | Cc1ncc(C[n+]2csc(CCO)c2C)c(N)n1.[O-][N+](=O)[O-] |
| IUPAC Name | 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;nitrate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?