missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Hydrochloride (White Powder), Fisher BioReagents

Product Code. 10113533
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10113533 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10113533 Supplier Fisher BioReagents Supplier No. BP892100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
This vitamin is suitable for use in systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-03-8
Melting Point 260°C
Color White
Density 1.4g/cm³
pH 2.7 to 3.4
Assay Percent Range ≥98 %
Packaging Glass Bottle
Molecular Formula C12H18Cl2N4OS
Quantity 100 g
Residue after Evaporation 0.2% max.
Merck Index 15, 9447
Synonym thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
SMILES CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
Molecular Weight (g/mol) 337.263
PubChem CID 6202
ChEBI CHEBI:49105
Formula Weight 300.66
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Thiamine Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.