missing translation for 'onlineSavingsMsg'
Learn More

Thiamine hydrochloride, MedChemExpress

Product Code. 16449126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449126 100 mg 100mg
16412549 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16449126 Supplier Medchem Express Supplier No. HYN0680
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Thiamine hydrochloride (Thiamine chloride hydrochloride) is an essential micronutrient needed as a cofactor for many central metabolic enzymes.

Thiamine hydrochloride (Thiamine chloride hydrochloride) is an essential micronutrient needed as a cofactor for many central metabolic enzymes.

Specifications

Chemical Name or Material Thiamine hydrochloride
CAS 67-03-8
Color White
Purity Grade Notes Research
Molecular Formula C12H18Cl2N4OS
Quantity 100 mg
Synonym Thiamine chloride hydrochlorideVitamin B1 hydrochloride
Solubility Information H2O : 100 mg/mL (296.50 mM; Need ultrasonic) ∣DMSO : 6.67 mg/mL (19.78 mM; Need ultrasonic)
SMILES CC1=C(CCO)SC=[N+]1CC2=CN=C(C)N=C2N.[Cl-].Cl
Molecular Weight (g/mol) 337.27
Formula Weight 337.27
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.